Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSIG4 Antibody (2174B), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB176284 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176284 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176284 Supplier Novus Biologicals Supplier No. FAB46461X
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

VSIG4 Monoclonal antibody specifically detects VSIG4 in Human samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen VSIG4
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 2174B
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived human VSIG4, Arg20-Pro283, Accession # Q9Y279
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11326
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.