Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSIG4 Rabbit anti-Mouse, DyLight 650, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB081480 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081480 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081480 Supplier Novus Biologicals Supplier No. NBP298230C

Rabbit Polyclonal Antibody

VSIG4 Polyclonal specifically detects VSIG4 in Mouse samples. It is validated for ELISA.

Specifications

Antigen VSIG4
Applications ELISA
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant VSIG4 (NP_808457.1; Met1-Pro187).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11326
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.