Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSTM2A Antibody (1024735) [FITC], Novus Biologicals™
SDP

Catalog No. NB402646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402646 Supplier Novus Biologicals Supplier No. FAB100371F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VSTM2A Monoclonal antibody specifically detects VSTM2A in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF

Specifications

Antigen VSTM2A
Applications Flow Cytometry, ELISA, CyTOF
Classification Monoclonal
Clone 1024735
Conjugate FITC
Dilution Flow Cytometry, ELISA, CyTOF-ready
Formulation PBS
Gene Alias MGC33530, V-set and transmembrane domain containing 2, V-set and transmembrane domain containing 2A, V-set and transmembrane domain-containing protein 2A, VSTM2
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived human VSTM2A protein, Ser25-Phe244, Accession # Q8TAG5-2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222008
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.