Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSTM2L Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB050311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050311 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050311 Supplier Novus Biologicals Supplier No. NBP297976UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VSTM2L Polyclonal specifically detects VSTM2L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen VSTM2L
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C20orf102, chromosome 20 open reading frame 102, dJ1118M15.2, V-set and transmembrane domain containing 2 like, V-set and transmembrane domain-containing protein 2-like protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human VSTM2L fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128434
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.