Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSTM4 Antibody (2694A), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB176012 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176012 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176012 Supplier Novus Biologicals Supplier No. FAB10767E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

VSTM4 Monoclonal specifically detects VSTM4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen VSTM4
Applications Flow Cytometry
Classification Monoclonal
Clone 2694A
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias FLJ31737, hypothetical protein LOC196740, V-set and transmembrane domain containing 4
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line CHO-derived human VSTM4, Leu24-Tyr180, Accession # Q8IW00-1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196740
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.