Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSTM5 Antibody (2696A), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB339888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339888 Supplier Novus Biologicals Supplier No. FAB10872MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

VSTM5 Monoclonal specifically detects VSTM5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen VSTM5
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 2696A
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias C11orf90, V-set and transmembrane domain containing 5, V-set and transmembrane domain-containing protein 5
Host Species Rabbit
Immunogen Human embryonic kidney cell HEK293-derived human VSTM5, Met1-His147, Accession # NP_001138343
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387804
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.