Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSV-G Epitope Tag Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB292334 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292334 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292334 Supplier Novus Biologicals Supplier No. NB1002484PE
Add to Cart
Add to Cart

Goat Polyclonal Antibody

VSV-G Epitope Tag Polyclonal specifically detects VSV-G Epitope Tag in Tag samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen VSV-G Epitope Tag
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias Vesicular stomatitis virus glycoprotein tag, VSV epitope tag, VSV tag, VSVG, VSV-G epitope tag, vsvg tag
Host Species Goat
Immunogen Synthetic peptide: YTDIEMNRLGK conjugated to KLH.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.