Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VTA1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB085564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB085564 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB085564 Supplier Novus Biologicals Supplier No. NBP299526AF532

Rabbit Polyclonal Antibody

VTA1 Polyclonal specifically detects VTA1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen VTA1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias C6orf55DRG1, Dopamine-responsive gene 1 protein, DRG-1, FLJ27228, homolog of mouse SKD1-binding protein 1, LIP5HSPC228, LYST-interacting protein 5, My012, SBP1chromosome 6 open reading frame 55, SKD1-binding protein 1, vacuolar protein sorting-associated protein VTA1 homolog, Vps20-associated 1 homolog (S. cerevisiae)
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human VTA1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 51534
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.