Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VTI1A Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342041 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342041 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342041 Supplier Novus Biologicals Supplier No. NBP281876MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VTI1A Polyclonal antibody specifically detects VTI1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen VTI1A
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias MVti1, vesicle transport through interaction with t-SNAREs homolog 1A, vesicle transport through interaction with t-SNAREs homolog 1A (yeast)
Host Species Rabbit
Immunogen VTI1a antibody was raised against a 19 amino acid peptide near the center of human VTI1a. The immunogen is located within amino acids 50 - 100 of VTI1a. Amino Acid Squence: REIPPQSRGMYSNRMRSYK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 143187
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.