Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VU 0255035
GREENER_CHOICE

Catalog No. p-7116043
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-272-5 25 mg
37-275 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-272-5 Supplier Tocris Bioscience™ Supplier No. 3727/25
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly selective M1 antagonist

VU 0255035 is a highly selective muscarinic M1 antagonist (Ki = 14.87 nM). Targets the M1 orthosteric site with greater than 75-fold selectivity over the other receptor subtypes. Reduces pilocarpine-induced seizures in mice. Brain penetrant.

Specifications

Chemical Name or Material N-[3-Oxo-3-[4-(4-pyridinyl)-1-piperazinyl]propyl]-2,1,3-benzothiadiazole-4-sulfonamide
CAS 1135243-19-4
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C18H20N6O3S2
Formula Weight Observed MW: 432.52
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.