Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VWF Recombinant Rabbit Monoclonal Antibody (8O6D7)
GREENER_CHOICE

Catalog No. PIMA535178
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535178 Supplier Invitrogen™ Supplier No. MA535178
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: APACQVTCQH PEPLACPVQC VEGCHAHCPP GKILDELLQT CVDPEDCPVC EVAGRRFASG KKVTLNPSDP EHCQICHCDV VNLTCEACQE PGGLVVPPTD APVSPTTLYV EDISEPPLHD FYCSRLLDLV FLLDGSSRLS EAEFEVLKAF V.

Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VWF
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 8O6D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene VWF
Gene Accession No. P04275, Q8CIZ8
Gene Alias 6820430P06Rik; AI551257; B130011O06Rik; C630030D09; Coagulation Factor VIII; coagulation factor VIII VWF; EMBL:CAB37852.1}; F8VWF; factor viii related antigen; von Willebrand antigen 2; von Willebrand antigen II; von Willebrand Disease (vWD); von Willebrand factor; Von Willebrand factor homolog; VWD; vwf; vwf {ECO:0000312
Gene Symbols VWF
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1150-1300 of human von Willebrand factor (VWF) (P04275).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22371, 7450
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.