Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VX 745
GREENER_CHOICESDP

Catalog No. 391510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-151-0 10 mg
39-155-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-151-0 Supplier Tocris Bioscience™ Supplier No. 3915/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective p38α inhibitor

VX 745 is a highly potent and selective p38α/MAPK14 inhibitor, also inhibits p38β (IC50 values are 10 nM and 220 nM respectively). VX 745 blocks TNFα production in LPS-stimulated HWB in vitro (IC50 = 51-180 nM). Also reduces phosphorylation of p38MAPKα and the downstream target Hsp27, leading to vasodilation, reduces IL-1β levels and amyloid plaques in cortex and hippocampus and lowers CSF tau levels in vivo.

Chemical Identifiers

CAS 209410-46-8
Molecular Formula C19H9Cl2F2N3OS

Specifications

Quantity 10 mg
Formula Weight 436.26
Purity >98%
Molecular Weight (g/mol) 436.26
Chemical Name or Material VX 745
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.