Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WAC Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB184346 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184346 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184346 Supplier Novus Biologicals Supplier No. NBP282057IR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WAC Polyclonal antibody specifically detects WAC in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen WAC
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias 1110067P07Rik, bA48B24, bA48B24.1, BM-016, FLJ31290, KIAA1844, MGC10753, PRO1741, WW domain containing adaptor with coiled-coil, WW domain-containing adapter protein with coiled-coil, WW domain-containing adaptor with coiled coil, Wwp4
Host Species Rabbit
Immunogen WAC antibody was raised against a 19 amino acid peptide near the center of human WAC . The immunogen is located within amino acids 210 - 260 of WAC. Amino Acid Squence: CILSQTSRHNDRDYRLPRAE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 51322
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.