Learn More
Walter Products™ Human Fetal Skull Model
SCI_ED
Catalog No. s07119 Shop All Walter Products Products
Click to view available options
:
Dimensions: 5.5 x 3.5 x 3.5 in. (14 x 9 x 9cm)
Shows characteristics of prenatal development during 30th week of pregnancy.
  • Visible fontanelle, which becomes bone over time
  • Allows students to study anatomy of fetal skull with ease
  • Mounted on durable base with movable jaw
  • Dimensions: 5.5 x 3.5 x 3.5 in. (14 x 9 x 9cm)
Specifications
For Use With (Application) Prenatal development during 30th week of pregnancy
Product Type Model, Human Fetal Skull
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.