Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WAPL Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB165245 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165245 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165245 Supplier Novus Biologicals Supplier No. NBP281779AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WAPL Polyclonal antibody specifically detects WAPL in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen WAPL
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias FOEKIAA0261friend of EBNA2 (Epstein-Barr virus nuclear protein 2), Friend of EBNA2 protein, WAPLwings apart-like protein homolog, wings apart-like homolog (Drosophila)
Host Species Rabbit
Immunogen WAPL antibody was raised against an 18 amino acid peptide near the amino terminus of human WAPL. The immunogen is located within amino acids 30 - 80 of WAPL. Amino Acid Squence: LGQKRPNFKPDIQEIPKK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 23063
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.