Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WAPL Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140080
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7245 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7245 Supplier Bethyl Laboratories, Inc Supplier No. A300268AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

WAPL (wings apart-like), also known as WAPAL or FOE, is a 190 amino acid protein that contains one WAPL domain and may play an important role in cell growth. It is expressed in an isoform dependent manner in heart, skeletal muscle (isoform 2) and uterine cervix tumor tissue (isoform 1). WAPL is involved in sister-chromatid adhesion and promotes release of cohesin from chromosomes by directly interacting with its regulatory subunits. WAPL is a new regulator of the development and metastasis of cancerous tissue.

Specifications

Antigen WAPL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene WAPL
Gene Accession No. Q7Z5K2
Gene Alias FOE, KIAA0261, WAPL, friend of EBNA2 (Epstein-Barr virus nuclear protein 2), friend of EBNA2 protein, wings apart-like protein homolog
Gene Symbols WAPL
Host Species Rabbit
Immunogen Between 1125 and 1175
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23063
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.