Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WASF1/WAVE1 Antibody, Novus Biologicals™
SDP

Catalog No. NB387862 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387862 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387862 Supplier Novus Biologicals Supplier No. NBP323363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WASF1/WAVE1 Polyclonal antibody specifically detects WASF1/WAVE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen WASF1/WAVE1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence 1:200
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias KIAA0269WAVEhomology of dictyostelium scar 1, Protein WAVE-1, SCAR1WASP family protein member 1, Verprolin homology domain-containing protein 1, WAS protein family, member 1, WAVE1FLJ31482, wisk
Host Species Rabbit
Immunogen A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal half of human WASF1/WAVE1. This sequence has high homology with similar regions in rat and mouse WASF1/WAVE1, and has less than 50% homology to similar regions in human WAVE2 and WAVE3. [UniProt# Q92558]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8936
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.