Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WASF3/WAVE3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220885 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220885 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220885 Supplier Novus Biologicals Supplier No. NBP220885
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WASF3/WAVE3 Polyclonal specifically detects WASF3/WAVE3 in Human samples. It is validated for Western Blot.

Specifications

Antigen WASF3/WAVE3
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UPY6
Gene Alias KIAA0900WASP family Verprolin-homologous protein 3, Protein WAVE-3, SCAR3WASP family protein member 3, Verprolin homology domain-containing protein 3, WAS protein family, member 3, WAVE3Brush-1, wisk
Gene Symbols WASF3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human WASF3. The exact sequence is proprietary.
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10810
Test Specificity Human. Species reactivity based on sequence identity: Mouse (84%), Cow (86%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.