Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Azlon™ Solvent Venting Wash Bottles

Catalog No. 03406155 Shop All Dynalab Corporation Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Closure Color:
Green
Red
White

Catalog No. 03-406-155

Supplier: Dynalon™ 5064740003

Only null left
Add to Cart
Add to Cart

Unique two-way valve stop vapour pressure build-up and eliminates the hazard of volatile solvent leaks

Translucent low-density polyethylene wash bottles feature a unique two-way valve that stops vapor pressure build-up and eliminates the hazard of volatile solvent leaks. The wide neck provides for easier and safer filling. Plus, for quick identification, cadmium-free polypropylene dispensing closures are color-coded. And, for optimum control in dispensing, the jet is precision molded. Each bottle features a clear, indelible label describing the contents, international hazard codes and instructional diagrams for safe usage. Unlabeled bottles with write-on label areas and a complete set of adhesive hazard labels for custom use are also available. Supplied in convenient shelf packs of five.

Order Info

Bottles are available in 250 or 500mL capacities with five common solvent labels.

Compliance

All bottles have SPI recycling code stamped on base.

Specifications

Capacity 1000mL (32oz)
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color White, Translucent
Diameter (English) 3.54 in.
Height (Metric) With Closure 273mm
Material LDPE
Packaging CA/5
Quantity CA/5
Sterility Non-sterile
Width (Metric) 90mm
Closure Color White
Height (Metric) 209 mm
Product Type Wash Bottle
Width (English) 3.54 in.
Height (English) 8.23 in.
Chemical Label Custom Write-on
Diameter (Metric) 90 mm
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.