Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, ACS Reagent Grade, ASTM Type I, Ricca Chemical
SDP

Catalog No. 915032
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
55 gal.
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

Color Colorless
Melting Point 0°C
CAS Min % 98.0
Boiling Point 100°C
CAS Max % 100.0
Conductivity max. 0.056
Synonym dihydrogen oxide, dihydrogen monoxide
Solubility Information Miscible
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
Formula Weight 18.01
Percent Purity 100%
Grade ACS
Packaging Natural Poly Bottle
Sterility Non-sterile
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water, ASTM Type I
Show More Show Less
ShelfLife 18 months
DOTInformation Not Regulated by DOT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.