Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water, Distilled, Reagent Grade, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
1 L
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Cubitainer
Poly Drum
Specifications
Specifications
| Color | Colorless |
| Conductivity | max. 1.5 μS/cm |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| IUPAC Name | oxidane |
| Molecular Weight (g/mol) | 18.015 |
| Percent Purity | 100% |
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
| Sterility | Non-sterile |
| Quantity | 1 L |
| Show More |
Safety and Handling
| ShelfLife | 18 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction