Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water, Reagent (Deionized water), ACS, Alfa Aesar™
Click to view available options
Quantity:
1L
4L
4 x 1L
Specifications
Appearance | Liquid |
Solubility Information | Miscible in methanol,acetone,Ethanol. |
Formula Weight | 18.02 |
Physical Form | Liquid |
Grade | ACS |
Odor | Odorless |
Melting Point | 0°C |
Density | 1 |
Color | Colorless |
Boiling Point | 100°C |
Show More |
Safety and Handling
EINECSNumber : 231-791-2
TSCA : Yes
Recommended Storage : Ambient temperatures