Learn More
Eisco™ Solar Furnace
SCI_ED
Catalog No. s12958 Shop All Eisco Products
Showcases the transfer of energy between solar radiation and thermal energy. The EISCO Solar Furnace is ideal for physics classroom demonstrations. Light from the Sun's rays are reflected off of a 30cm (11.8 in.) diameter parabolic reflector and are focused onto a black colored copper cup.
  • Ideal for classroom demonstration on solar energy
  • 11.8 in. parabolic mirror to focus the light rays
  • Sun's rays effectively heat the copper cup at end of column
Specifications
Product Type Solar Furnace
For Use With (Application) Classroom demonstration on solar energy
Includes Parabolic reflector, Black colored copper cup, Stand
Diameter (English) 11.8 in.
Height (English) 18 in.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.