Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WAY 161503 hydrochloride
GREENER_CHOICE

Catalog No. 180110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-011-0 10 mg
18-015-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-011-0 Supplier Tocris Bioscience™ Supplier No. 1801/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective 5-HT2C agonist

Potent and selective 5-HT2C receptor agonist (Ki = 4 nM; EC50 = 12 nM). Antidepressant following systemic administration in vivo.

Chemical Identifiers

CAS 276695-22-8
Molecular Formula C11H12Cl3N3O

Specifications

Quantity 10 mg
Formula Weight 308.59
Purity >99%
Molecular Weight (g/mol) 308.587
Chemical Name or Material WAY 161503 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.