Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WBSCR22 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $610.00
Specifications
Antigen | WBSCR22 |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
WBSCR22 Polyclonal specifically detects WBSCR22 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
WBSCR22 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
114049 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: ELFYDETEARKYVRNSRMIDIQTRMAGRALELLYLPENKPCYLLDIGCGTGLSGSYLSDEGHYWVGLDISPAMLDE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
Polyclonal | |
Rabbit | |
Human | |
EC 2.1.1.-, FLJ44236, HASJ4442, HUSSY-3, MGC19709, MGC2022, MGC5140, PP3381, WBMT, Williams Beuren syndrome chromosome region 22, Williams Beuren syndrome chromosome region 22 protein, Williams-Beuren candidate region putative methyltransferase, Williams-Beuren syndrome chromosomal region 22 protein | |
WBSCR22 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title