Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WC1(+) gamma delta T cell Antibody (B7A1), Novus Biologicals™
SDP

Catalog No. NBP261043 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261043 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP261043 Supplier Novus Biologicals Supplier No. NBP261043
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WC1(+) gamma delta T cell Monoclonal specifically detects WC1(+) gamma delta T cell in Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen WC1(+) gamma delta T cell
Applications Flow Cytometry, Immunohistochemistry, Functional Assay
Classification Monoclonal
Clone B7A1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000
Formulation Ascites with 0.09% Sodium Azide
Host Species Mouse
Immunogen Bovine leukocytes
Purification Method Unpurified
Quantity 0.1 mg
Primary or Secondary Primary
Target Species Goat, Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.