Learn More
Description
Specifications
Specifications
| Antigen | WDR11 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate |
| Formulation | TBS and 0.1% BSA with 0.09% Sodium Azide |
| Gene Accession No. | Q9BZH6 |
| Gene Alias | Bromodomain and WD repeat-containing protein 2, BRWD2DKFZp434L1715, DR11, FLJ10506, FLJ42531, KIAA1351bromodomain and WD repeat domain containing 2, WD repeat domain 11, WD repeat domain 15, WD repeat-containing protein 11, WD repeat-containing protein 15, WDR15 |
| Gene Symbols | WDR11 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
