Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR42A Antibody, Novus Biologicals™
SDP

Catalog No. NB10077300 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10077300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10077300 Supplier Novus Biologicals Supplier No. NB10077300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDR42A Polyclonal specifically detects WDR42A in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen WDR42A
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 1:10-1:500
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_056541.2
Gene Alias DDB1 and CUL4 associated factor 8, DDB1- and CUL4-associated factor 8, DKFZp781G1096, FLJ35857, H326MGC117276, MGC118891, MGC99640, WD repeat domain 42A, WD repeat-containing protein 42A, WDR42A
Gene Symbols DCAF8
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 500 and 550 of human WD repeat domain 42A using the numbering given in entry NP_056541.2 (GeneID 50717).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50717
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.