Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR45 Antibody, Novus Biologicals™
SDP

Catalog No. NB171879 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171879 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171879 Supplier Novus Biologicals Supplier No. NBP318255
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WDR45 Polyclonal antibody specifically detects WDR45 in Human, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen WDR45
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100
Formulation PBS, 50% glycerol
Gene Alias JM5, WD repeat domain 45, WD repeat domain phosphoinositide-interacting protein 4, WD repeat-containing protein 45, WD45 repeat protein interacting with phosphoinositides 4, WDRX1, WDRXI4, WIPI-4, X-linked 1
Host Species Rabbit
Immunogen Synthetic peptide from the C-terminal of Human WDR45(aa. 312-322)
Purification Method Peptide affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 11152
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.