Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR5 Rabbit anti-Human, Rat, Clone: 3S6R9, Novus Biologicals™
SDP

Catalog No. NB167415 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167415 100 μg
NB167416 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167415 Supplier Novus Biologicals Supplier No. NBP316162100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

WDR5 Monoclonal antibody specifically detects WDR5 in Human, Rat samples. It is validated for Western Blot

Specifications

Antigen WDR5
Applications Western Blot
Classification Monoclonal
Clone 3S6R9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BIG3, BIG-3, BMP2-induced 3-kb gene protein, SWD3, SWD3, Set1c WD40 repeat protein, homolog, WD repeat domain 5, WD repeat-containing protein 5, WD-repeat protein 5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 235-334 of human WDR5 (P61964).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 11091
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.