Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR5 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB170502 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170502 100 μg
NB170503 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170502 Supplier Novus Biologicals Supplier No. NBP318672100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WDR5 Polyclonal antibody specifically detects WDR5 in Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen WDR5
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100-1:500
Formulation Whole antiserum
Gene Accession No. P61964
Gene Alias BIG3, BIG-3, BMP2-induced 3-kb gene protein, SWD3, SWD3, Set1c WD40 repeat protein, homolog, WD repeat domain 5, WD repeat-containing protein 5, WD-repeat protein 5
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against mouse Wdr5 (WD (tryptophan-aspartate) repeat domain protein 5), using two KLH-conjugated synthetic peptides containing an amino acid sequence from the central part of the protein (1).
Purification Method Whole Antiserum
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 11091
Target Species Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.