Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR51B Antibody, Novus Biologicals™
SDP

Catalog No. NBP15630720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15630720 20 μL
NBP156307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15630720 Supplier Novus Biologicals Supplier No. NBP15630720UL

Rabbit Polyclonal Antibody

WDR51B Polyclonal specifically detects WDR51B in Human samples. It is validated for Western Blot.

Specifications

Antigen WDR51B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8TC44
Gene Alias FLJ14923, FLJ41111, PIX1, POC1 centriolar protein homolog B, POC1 centriolar protein homolog B (Chlamydomonas), TUWD12, WD repeat domain 51B, WD repeat-containing protein 51B, WDR51B
Gene Symbols POC1B
Host Species Rabbit
Immunogen Synthetic peptides corresponding to WDR51B(WD repeat domain 51B) The peptide sequence was selected from the N terminal of WDR51B. Peptide sequence GNLLASASRDRTVRLWIPDKRGKFSEFKAHTAPVRSVDFSADGQFLATAS.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282809
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.