Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR61 Antibody, Novus Biologicals™
SDP

Catalog No. NBP259231 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259231 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259231 Supplier Novus Biologicals Supplier No. NBP259231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDR61 Polyclonal specifically detects WDR61 in Human, Drosophila samples. It is validated for Western Blot, Chromatin Immunoprecipitation.

Specifications

Antigen WDR61
Applications Western Blot, ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 1-5 ug
Formulation PBS with 0.05% Sodium Azide and 0.05% ProClin 300
Gene Alias Meiotic recombination REC14 protein homolog, REC14, recombination protein REC14, SKI8, WD repeat domain 61, WD repeat-containing protein 61
Gene Symbols WDR61
Host Species Rabbit
Immunogen The exact immunogen is propietary information.
Purification Method Protein G purified
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80349
Target Species Human, Drosophila
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.