Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR61 Antibody (OTI4C4), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP274866R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274866R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274866R Supplier Novus Biologicals Supplier No. NBP274866R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WDR61 Monoclonal specifically detects WDR61 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen WDR61
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4C4
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Meiotic recombination REC14 protein homolog, REC14, recombination protein REC14, SKI8, WD repeat domain 61, WD repeat-containing protein 61
Gene Symbols WDR61
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 92-305 of human WDR61 (NP_079510) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 80349.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.