Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR83OS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159969
Description
WDR83OS Polyclonal specifically detects WDR83OS in Human samples. It is validated for Western Blot.Specifications
C19orf56 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
C19orf56, chromosome 19 open reading frame 56, hypothetical protein LOC51398, PTD008, WD repeat domain 83 opposite strand | |
Rabbit | |
Affinity purified | |
RUO | |
51398 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9Y284 | |
WDR83OS | |
Synthetic peptides corresponding to C19ORF56 The peptide sequence was selected from the N terminal of C19ORF56. Peptide sequence STNNMSDPRRPNKVLRYKPPPSECNPALDDPTPDYMNLLGMIFSMCGLML. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Equine: 100%; Pig: 100%; Rabbit: 100%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction