Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR89 Antibody, Novus Biologicals™
SDP

Catalog No. NBP103335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP103335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP103335 Supplier Novus Biologicals Supplier No. NBP103335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDR89 Polyclonal specifically detects WDR89 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen WDR89
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 1:10-1:500
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_542397.1
Gene Alias C14orf150, MGC9907, WD repeat domain 89, WD repeat-containing protein 89
Gene Symbols WDR89
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 337 and 387 of human WD repeat domain 89 using the numbering given in entry NP_542397.1 (GeneID 112840).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112840
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.