Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR90 Antibody, Novus Biologicals™
SDP

Catalog No. NB397552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397552 25 μL
NBP231888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397552 Supplier Novus Biologicals Supplier No. NBP23188825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

WDR90 Polyclonal specifically detects WDR90 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen WDR90
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q96KV7
Gene Alias C16orf15, C16orf16, C16orf17, C16orf18, C16orf19, chromosome 16 open reading frame 15, chromosome 16 open reading frame 17, chromosome 16 open reading frame 18, chromosome 16 open reading frame 19, FLJ36483, FLJ44660, KIAA1924chromosome 16 open reading frame 16, WD repeat domain 90, WD repeat-containing protein 90
Gene Symbols WDR90
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: FKEFKSTATWLQFPLVLEARTPQRDLVGLAPSGARWTCLQLDLQDVLLVYLNRCYGHLKSIRLCASLLVRNLYTSDLCFEPAISGAQWAKLPVT
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197335
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.