Learn More
American Educational Products Weather Wall Charts
Shop All American Educational Products LLC Products

Description
Record and compare the day's or school week's weather forecast to actual weather conditions. Using static cling weather symbols (sold separately, S79056), students track weather and learn more about weather patterns. Complements the Weather Wall Classroom Map (S79059).
Specifications
Specifications
| Product Type | School Days Forecast Wall Chart |
| For Use With (Application) | Weather system clings, teach record keeping skills, used on the “weather wall” to show the weather for the day of the week |
| Grade | K to 4 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.