Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Contains 1mL each of these mixtures.
30096: Unleaded Gasoline (ASTM Class 2 Accelerant)
- Unleaded gasoline: unweathered (8006-61-9)
30097: Unleaded Gasoline (ASTM Class 2 Accelerant)
- Unleaded gasoline: 25% weathered (8006-61-9)
30098: Unleaded Gasoline (ASTM Class 2 Accelerant)
- Unleaded gasoline: 50% weathered (8006-61-9)
30099: Unleaded Gasoline (ASTM Class 2 Accelerant)
- Unleaded gasoline: 75% weathered (8006-61-9)
30436: Unleaded Gasoline (ASTM Class 2 Accelerant)
- Unleaded gasoline: 99% weathered (8006-61-9)
Specifications
Specifications
Description | 0%, 25%, 50%, 75%, 99% Weathered |
Analyte or Component Names | Contains 1 mL each of these mixtures: Unleaded Gasoline Standard; Unleaded Gasoline Standard: 25% Weathered; Unleaded Gasoline Standard: 50% Weathered; Unleaded Gasoline Standard: 75% Weathered; Unleaded Gasoline Standard: 99% Weathered |
Type | Multi-component Standard |
Element | Unleaded Gasoline Standard, Unleaded Gasoline Standard: 25% Weathered, Unleaded Gasoline Standard: 50% Weathered, Unleaded Gasoline Standard: 75% Weathered, Unleaded Gasoline Standard: 99% Weathered |
Quantity | 1 mL |
Safety and Handling
ShelfLife | 6 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction