Learn More
Labconco™ Welch Vacuum Pump
SDP
Catalog No. p-200112411 Shop All Labconco Corporation Products
Click to view available options
Voltage:
115 V
220/240 V
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
This chemically resistant vacuum pump is used for lyophilization or concentration of samples containing harsh chemicals like TFA, Acetonitrile, etc.
  • This chemically resistant vacuum pump is used for lyophilization or concentration of samples containing harsh chemicals like TFA, Acetonitrile, etc.
  • An alumina filter element neutralizes acids and removes reaction products.
  • Internal oil case is PTFE coated.
Specifications
Flow Rate 133 L/min
Pump Type Rotary Vane Vacuum Pump
Enclosure Rating IP54
Maximum Pressure 0.002 mBar
Voltage 220/240 V
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.