Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Economy Polypropylene Lab Coat

Catalog No. 191301293F Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Standard weight polypropylene lab coat protects against dry particulates and worn when full coverage is not necessary.

  • Standard weight spunbond polypropylene
  • Chest and hip pockets
  • Snap buttons for quick donning
  • Professional collar
  • Single-use, white

Specifications

Product Type General Purpose Lab Coat
Size 3X-Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
No. of Pockets 2
Model 3514
Seam Type Serged
For Use With (Application) Manufacturing, Pharmaceuticals, Cleaning, Laboratories
Construction Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.