Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Nile Virus C Protein Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB221535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB221535 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB221535 Supplier Novus Biologicals Supplier No. NBP241054AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

West Nile Virus C Protein Polyclonal antibody specifically detects West Nile Virus C Protein in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen West Nile Virus C Protein
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Capside protein C, Core protein, West Nile Core, West Nile Virus Core protein, WNV core
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the carboxy terminus of the West Nile Virus core protein. The immunogen is located within amino acids 80 - 130 of West Nile Virus Core. Amino Acid Squence: SAINRRSTKQKKRGG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 912267
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.