Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Nile Virus Envelope Antibody (03), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB351852 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351852 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351852 Supplier Novus Biologicals Supplier No. NBP306462MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

West Nile Virus Envelope Monoclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for ELISA

Specifications

Antigen West Nile Virus Envelope
Applications ELISA
Classification Monoclonal
Clone 03
Conjugate mFluor Violet 450 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant West Nile Virus Envelope (Accession#: ABA62343.1; Gln586-Ala705).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.