Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Nile Virus Envelope Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB355483 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355483 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355483 Supplier Novus Biologicals Supplier No. NBP241055MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

West Nile Virus Envelope Polyclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for ELISA

Specifications

Antigen West Nile Virus Envelope
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the center of the West Nile Virus Envelope protein. The immunogen is located within amino acids 500 - 550 of West Nile Virus Envelope Protein. Amino Acid Squence: GSTTWRNRETLMEFE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.