Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
West Nile Virus Mouse anti-Virus, Unconjugated, Clone: E16, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | West Nile Virus |
---|---|
Clone | E16 |
Applications | Immunocytochemistry, Neutralization, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF22342MI
|
MilliporeSigma
MABF223425UG |
25 μg |
Each for $182.31
|
|
|||||
MABF22341MI
|
MilliporeSigma
MABF2234100UG |
100 μg |
Each for $457.74
|
|
|||||
Description
West Nile Virus Monoclonal antibody specifically detects West Nile Virus in Virus samples. It is validated for Immunocytochemistry, Neutralizing, Western Blotting.Specifications
West Nile Virus | |
Immunocytochemistry, Neutralization, Western Blot | |
Unconjugated | |
Mouse | |
Virus | |
E | |
IgG2b κ | |
Protein G Purified | |
Clone E16 is a mouse monoclonal antibody that specifically detects envelope protein of West Nile Virus. It targets an epitope within the DIII lateral ridge region. |
E16 | |
Monoclonal | |
Purified | |
Epigenetics and Nuclear Function | |
E protein;Envelope protein;Env | |
West Nile Virus envelope protein. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
∼50 kDa observed; 31.88 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title