Learn More
Description
Specifications
Specifications
| Antigen | West Nile Virus MP |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | West Nile Virus envelope glycoprotein M |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acid residues 8-27 (GESTLANKKGAWLDSTKATR) of West Nile Virus MP. Genbank accession no. NP_776013. |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
