Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Nile Virus MP Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10056743 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056743 0.1 mg
NB10056743T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056743 Supplier Novus Biologicals Supplier No. NB10056743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 11 publications

West Nile Virus MP Polyclonal specifically detects West Nile Virus MP in Hamster, Human, Mouse, Rhesus Macaque, Virus samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen West Nile Virus MP
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:20-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:1000. Use reported in scientific literature (Tan et al (2009)), Immunoprecipitation reported in scientific literature (Tan et al (2009)), Immunohistochemistry-Paraffin 1.0 ug/ ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias West Nile Virus envelope glycoprotein M
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acid residues 8-27 (GESTLANKKGAWLDSTKATR) of West Nile Virus MP. Genbank accession no. NP_776013.
Purification Method Affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity West Nile Virus glycoprotein M (Genbank accession no. NP_776013).
Target Species Hamster, Mouse, Human, Mouse, Rhesus Macaque, Virus
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.