Learn More
Description
Specifications
Specifications
| Antigen | West Nile Virus MP |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:20-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:1000. Use reported in scientific literature (Tan et al (2009)), Immunoprecipitation reported in scientific literature (Tan et al (2009)), Immunohistochemistry-Paraffin 1.0 ug/ ml |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | West Nile Virus envelope glycoprotein M |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acid residues 8-27 (GESTLANKKGAWLDSTKATR) of West Nile Virus MP. Genbank accession no. NP_776013. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
