Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Nile Virus MP Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB219752 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB219752 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB219752 Supplier Novus Biologicals Supplier No. NBP241053V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

West Nile Virus MP Polyclonal antibody specifically detects West Nile Virus MP in Virus samples. It is validated for ELISA

Specifications

Antigen West Nile Virus MP
Applications ELISA
Classification Polyclonal
Conjugate DyLight 405
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the carboxy terminus of the West Nile Virus MP. The immunogen is located within amino acids 220 - 270. Amino Acid Squence: LDSTKATRYLVKTES
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.