Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WFDC5 Antibody (M47-P2D5), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309539 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309539 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309539 Supplier Novus Biologicals Supplier No. NBP250363MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WFDC5 Monoclonal antibody specifically detects WFDC5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen WFDC5
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone M47-P2D5
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias dJ211D12.5, PRG5protease inhibitor WAP1, Putative protease inhibitor WAP1, WAP four-disulfide core domain 5, WAP four-disulfide core domain protein 5, WAP1p53-responsive gene 5 protein
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide TERVHDGRPG
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Signaling
Primary or Secondary Primary
Gene ID (Entrez) 149708
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.